Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027323.2 | NP_081599.2 | 1 - 2 | 161 | 72 | |
GenBank mRNA | AK012640.1 | - | 1 - 2 | 126 | 72 | |
AV513732.1 | - | 1 - 2 | 159 | 72 | ||
BC107023.2 | - | 1 - 2 | 139 | 72 | ||
BC125007.1 | - | 1 - 2 | 144 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |