Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rimbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081388.1 | NP_001074857.1 | 18 - 19 | 3502 | 84 | |
GenBank mRNA | AK038814.1 | - | 17 - 18 | 2970 | 84 | |
AK044685.1 | - | 17 - 18 | 3602 | 84 | ||
AK122245.2 | - | 9 - 10 | 2315 | 84 | ||
AK147417.1 | - | 14 - 15 | 2908 | 84 | ||
BC151100.1 | - | 14 - 15 | 3093 | 9088013 | 84 | |
BC151101.1 | - | 14 - 15 | 3093 | 9088014 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |