Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rimbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081388.1 | NP_001074857.1 | 2 - 3 | 482 | 77 | |
GenBank mRNA | AK044685.1 | - | 2 - 3 | 603 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons |