Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029749.2 | NP_084025.2 | 13 - 14 | 2224 | 86 | |
GenBank mRNA | AK041514.1 | - | 1 - 2 | 420 | 86 | |
AY336567.1 | - | 12 - 13 | 2097 | 86 | ||
BC137852.1 | - | 12 - 13 | 2117 | 8860871 | 86 | |
BM963346.1 | - | 1 - 2 | 509 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |