Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164833.1 | NP_001158305.1 | 2 - 3 | 392 | 67 | |
NM_001164834.1 | NP_001158306.1 | 2 - 3 | 392 | 67 | ||
NM_153400.4 | NP_700449.2 | 2 - 3 | 392 | 67 | ||
GenBank mRNA | AB094663.1 | - | 2 - 3 | 312 | 67 | |
AB094664.1 | - | 2 - 3 | 312 | 67 | ||
AK141196.1 | - | 2 - 3 | 351 | 67 | ||
AK158334.1 | - | 2 - 3 | 370 | 67 | ||
AY044240.2 | - | 2 - 3 | 568 | 67 | ||
BC106963.1 | - | 2 - 3 | 199 | 67 | ||
BC106964.1 | - | 2 - 3 | 199 | 40039097 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |