Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164833.1 | NP_001158305.1 | 9 - 10 | 1083 | 72 | |
NM_001164834.1 | NP_001158306.1 | 9 - 10 | 1083 | 72 | ||
NM_153400.4 | NP_700449.2 | 9 - 10 | 1083 | 72 | ||
GenBank mRNA | AB094663.1 | - | 9 - 10 | 1003 | 72 | |
AB094664.1 | - | 9 - 10 | 1003 | 72 | ||
AK141196.1 | - | 9 - 10 | 1042 | 72 | ||
AY044240.2 | - | 9 - 10 | 1259 | 72 | ||
BC106964.1 | - | 9 - 10 | 890 | 40039097 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |