Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dkkl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015789.3 | NP_056604.2 | 3 - 4 | 441 | 85 | |
GenBank mRNA | AF177399.1 | - | 3 - 4 | 346 | 85 | |
AF274312.1 | - | 3 - 4 | 400 | 85 | ||
AK131694.1 | - | 3 - 4 | 386 | 85 | ||
BC002215.1 | - | 3 - 4 | 419 | 3490482 | 85 | |
CA466481.1 | - | 3 - 4 | 444 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |