Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2116818 |
BC100441.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2116818 |
BC125429.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2116818 |
BC137644.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2116818 |
NM_029408.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2116818 |
AK017032.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P115655 |
AK076996.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P115655 |
NM_029408.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P115655 |
AK017032.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P115655 |
AK076996.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P115655 |
NM_029408.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P115655 |
AK017032.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
148766 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
89909 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
342261 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
12422 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
442743 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
106289 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
841041 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
603104 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
212919 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
571306 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
196665 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776960 |
AK003017.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776960 |
AK004036.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776960 |
AK005226.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776960 |
AK144665.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776960 |
BC029053.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776960 |
NM_133781.4 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
718339 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
34629 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
14079 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
30161 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P173703 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P106566 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P115655 |
AK076996.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P115655 |
NM_029408.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P115655 |
AK017032.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P115655 |
AK076996.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P115655 |
NM_029408.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P115655 |
AK017032.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2116818 |
BC100441.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2116818 |
BC125429.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2116818 |
BC137644.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2116818 |
NM_029408.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2116818 |
AK017032.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24039681 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24039664 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24039677 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24039675 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24039662 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24039667 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24039679 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24039671 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24039672 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24039669 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05719508 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05719519 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05719500 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01204158 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P106566 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P173703 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P115656 |
BC125429.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P115656 |
BC137644.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P115656 |
NM_029408.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P115656 |
AK017032.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P115655 |
AK076996.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P115655 |
NM_029408.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P115655 |
AK017032.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P115655 |
AK076996.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P115655 |
NM_029408.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P115655 |
AK017032.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P119492 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P173703 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P05719500 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P106566 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04313866 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039826 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039725 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039807 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039776 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039689 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039750 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039785 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039814 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039692 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039831 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039717 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039671 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039784 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039662 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039699 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039738 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039740 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039791 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039714 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039679 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039812 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039696 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039734 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039804 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039770 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039754 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039792 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039718 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039762 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039727 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039790 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039800 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039700 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039765 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039661 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039703 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039686 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039827 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039744 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039802 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039759 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039701 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039780 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039811 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039719 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039659 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039787 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039688 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039779 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039828 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039669 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039716 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039782 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039697 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039742 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039753 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039756 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039729 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039774 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039663 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039794 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039681 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039723 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039687 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039830 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039665 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039798 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039705 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039715 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039733 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039816 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24039786 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039661 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039695 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039756 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04521792 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039667 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039686 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039767 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039689 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039769 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039761 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04802625 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039765 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04521803 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039665 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039682 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039675 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04555759 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039699 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039753 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039671 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039770 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039679 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039754 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04313866 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039673 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039692 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039697 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039696 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039762 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039687 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04521801 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04944409 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24039669 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04555764 |
- |
- |