Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC025824.1 | - | 1 - 2 | 212 | 5100719 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |