Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500012821.mm.1 |
NM_001033219.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012821.mm.1 |
NM_001033219.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012821.mm.1 |
TC1500001698.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012813.mm.1 |
NM_001033219.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012813.mm.1 |
NM_001033219.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012813.mm.1 |
NM_001168255.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012813.mm.1 |
NM_001168255.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012813.mm.1 |
TC1500001698.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012816.mm.1 |
NM_001033219.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012816.mm.1 |
NM_001033219.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012816.mm.1 |
TC1500001698.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012817.mm.1 |
NM_001033219.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012817.mm.1 |
NM_001033219.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012817.mm.1 |
TC1500001698.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012819.mm.1 |
NM_001033219.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012819.mm.1 |
NM_001033219.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012819.mm.1 |
TC1500001698.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012818.mm.1 |
NM_001033219.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012818.mm.1 |
NM_001033219.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012818.mm.1 |
TC1500001698.mm.1 |
MTA TC |