Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016698.2 | NP_057907.2 | 5 - 6 | 1305 | 101 | |
GenBank mRNA | AB026621.1 | - | 5 - 6 | 1304 | 101 | |
AK004579.1 | - | 5 - 6 | 1299 | 102 | ||
AK034953.1 | - | 5 - 6 | 1304 | 101 | ||
AK129106.1 | - | 5 - 6 | 1293 | 101 | ||
AK146728.1 | - | 5 - 6 | 1305 | 101 | ||
AK149972.1 | - | 5 - 6 | 1306 | 101 | ||
AK156502.1 | - | 5 - 6 | 1302 | 101 | ||
AK165617.1 | - | 5 - 6 | 1305 | 101 | ||
AK168523.1 | - | 5 - 6 | 1300 | 101 | ||
AK172065.1 | - | 5 - 6 | 1303 | 101 | ||
AK178308.1 | - | 1 - 2 | 123 | 101 | ||
AK183331.1 | - | 1 - 2 | 124 | 101 | ||
AK207783.1 | - | 2 - 3 | 198 | 101 | ||
BC010342.1 | - | 5 - 6 | 1014 | 3602969 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |