Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sppl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029012.2 | NP_083288.2 | 5 - 6 | 454 | 76 | |
GenBank mRNA | AK014709.1 | - | 5 - 6 | 792 | 76 | |
BC023131.1 | - | 1 - 2 | 69 | 76 | ||
BC086646.1 | - | 5 - 6 | 819 | 6817386 | 76 | |
BU613738.1 | - | 5 - 6 | 567 | 76 | ||
CD806412.1 | - | 1 - 2 | 71 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |