Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026718.2 | NP_080994.2 | 5 - 6 | 633 | 66 | |
GenBank mRNA | AK039432.1 | - | 5 - 6 | 828 | 66 | |
AK156539.1 | - | 5 - 6 | 826 | 66 | ||
BC027765.1 | - | 3 - 4 | 305 | 66 | ||
BC049187.1 | - | 5 - 6 | 812 | 66 | ||
BC061692.1 | - | 5 - 6 | 633 | 6308996 | 66 | |
BC108413.1 | - | 5 - 6 | 1108 | 6487251 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |