Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005247.2 | NP_001005247.2 | 21 - 22 | 3552 | 87 | |
NM_001005248.2 | NP_001005248.2 | 21 - 22 | 3528 | 87 | ||
NM_001167864.1 | NP_001161336.1 | 21 - 22 | 3453 | 87 | ||
GenBank mRNA | AF534396.1 | - | 21 - 22 | 3349 | 87 | |
AF534397.1 | - | 21 - 22 | 3325 | 87 | ||
AF534398.1 | - | 19 - 20 | 2914 | 87 | ||
AF534399.1 | - | 22 - 23 | 3480 | 87 | ||
AK129265.1 | - | 21 - 22 | 3475 | 87 | ||
AK140780.1 | - | 10 - 11 | 1734 | 87 | ||
AK141088.1 | - | 20 - 21 | 4177 | 87 | ||
AK154612.1 | - | 21 - 22 | 3282 | 87 | ||
AK161039.1 | - | 12 - 13 | 1923 | 87 | ||
BC046405.1 | - | 16 - 17 | 2531 | 87 | ||
BC046443.1 | - | 21 - 22 | 3166 | 87 | ||
BC082542.1 | - | 21 - 22 | 3453 | 6405811 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |