Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146013.1 | NP_001139485.1 | 1 - 2 | 87 | 68 | |
GenBank mRNA | AB118159.1 | - | 1 - 2 | 34 | 68 | |
AY147186.2 | - | 1 - 2 | 240 | 68 | ||
BC157997.1 | - | 1 - 2 | 87 | 9007308 | 68 | |
BC172125.1 | - | 1 - 2 | 87 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |