Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243052.1 | NP_001229981.1 | 1 - 2 | 275 | 88 | |
GenBank mRNA | AK160159.1 | - | 1 - 2 | 276 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |