Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243052.1 | NP_001229981.1 | 6 - 7 | 98 | ||
NM_001243053.1 | NP_001229982.1 | 7 - 8 | 98 | |||
NM_009737.3 | NP_033867.1 | 6 - 7 | 98 | |||
GenBank mRNA | AF031467.1 | - | 6 - 7 | 98 | ||
AK137049.1 | - | 7 - 8 | 98 | |||
AK145300.1 | - | 6 - 7 | 98 | |||
AK160159.1 | - | 6 - 7 | 98 | |||
BC017688.1 | - | 6 - 7 | 4242224 | 98 | ||
BC048072.1 | - | 6 - 7 | 4166264 | 98 | ||
U68526.1 | - | 6 - 7 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |