Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900004745.mm.1 |
NM_010615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004745.mm.1 |
NM_010615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004745.mm.1 |
TC1900000529.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900004746.mm.1 |
NM_010615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004746.mm.1 |
NM_010615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004746.mm.1 |
TC1900000529.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900004747.mm.1 |
NM_010615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004747.mm.1 |
NM_010615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004747.mm.1 |
TC1900000529.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900004748.mm.1 |
NM_010615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004748.mm.1 |
NM_010615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004748.mm.1 |
TC1900000529.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900004742.mm.1 |
NM_010615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004742.mm.1 |
NM_010615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004742.mm.1 |
TC1900000529.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900004744.mm.1 |
NM_010615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004744.mm.1 |
NM_010615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004744.mm.1 |
TC1900000529.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900004743.mm.1 |
NM_010615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004743.mm.1 |
NM_010615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004743.mm.1 |
TC1900000529.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900004750.mm.1 |
NM_010615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004750.mm.1 |
NM_010615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004750.mm.1 |
TC1900000529.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900004749.mm.1 |
NM_010615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004749.mm.1 |
NM_010615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900004749.mm.1 |
TC1900000529.mm.1 |
MTA TC |