Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900012622.mm.1 |
NM_198300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012622.mm.1 |
TC1900001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012626.mm.1 |
NM_198300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012626.mm.1 |
TC1900001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012610.mm.1 |
NM_198300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012610.mm.1 |
TC1900001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012623.mm.1 |
NM_198300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012623.mm.1 |
TC1900001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012624.mm.1 |
NM_198300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012624.mm.1 |
TC1900001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012625.mm.1 |
NM_198300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012625.mm.1 |
TC1900001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012627.mm.1 |
NM_198300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012627.mm.1 |
TC1900001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012637.mm.1 |
NM_198300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012637.mm.1 |
TC1900001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012618.mm.1 |
NM_198300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012618.mm.1 |
TC1900001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012611.mm.1 |
NM_198300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012611.mm.1 |
TC1900001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012621.mm.1 |
NM_198300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012621.mm.1 |
TC1900001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012612.mm.1 |
NM_198300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012612.mm.1 |
TC1900001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012620.mm.1 |
NM_198300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012620.mm.1 |
TC1900001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012609.mm.1 |
NM_198300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012609.mm.1 |
TC1900001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012613.mm.1 |
NM_198300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012613.mm.1 |
TC1900001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012615.mm.1 |
NM_198300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012615.mm.1 |
TC1900001448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012617.mm.1 |
NM_198300.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012617.mm.1 |
TC1900001448.mm.1 |
MTA TC |