Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145831.3 | NP_665830.1 | 2 - 3 | 909 | 110 | |
GenBank mRNA | AF539811.1 | - | 2 - 3 | 909 | 110 | |
AK044891.1 | - | 2 - 3 | 910 | 110 | ||
AK140337.1 | - | 2 - 3 | 901 | 110 | ||
AK140347.1 | - | 2 - 3 | 904 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |