Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prune2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181348.4 | NP_851993.3 | 1 - 2 | 170 | 126 | |
GenBank mRNA | AK020727.1 | - | 1 - 2 | 171 | 126 | |
AK039241.1 | - | 1 - 2 | 171 | 126 | ||
AK165913.1 | - | 1 - 2 | 171 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mus musculus 1700 Probe spans exons |