Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF086810.1 | - | 1 - 2 | 54 | 77 | |
AF234838.1 | - | 6 - 7 | 1442 | 77 | ||
AK140319.1 | - | 6 - 7 | 1408 | 77 | ||
AK140325.1 | - | 6 - 7 | 1408 | 77 | ||
BC057334.1 | - | 6 - 7 | 1460 | 6811850 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |