Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011962.3 | NP_036092.1 | 5 - 6 | 1291 | 105 | |
GenBank mRNA | AF046783.1 | - | 5 - 6 | 699 | 105 | |
AK013195.1 | - | 5 - 6 | 837 | 105 | ||
AK033360.1 | - | 5 - 6 | 836 | 105 | ||
AK053320.1 | - | 5 - 6 | 933 | 105 | ||
AK088948.1 | - | 5 - 6 | 708 | 105 | ||
AK157741.1 | - | 5 - 6 | 800 | 105 | ||
BC043047.1 | - | 5 - 6 | 1290 | 5703967 | 105 | |
BC054734.1 | - | 5 - 6 | 1291 | 5720686 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |