Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piwil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021311.3 | NP_067286.1 | 1 - 2 | 104 | 95 | |
GenBank mRNA | AB032604.1 | - | 1 - 2 | 69 | 95 | |
AF438405.1 | - | 1 - 2 | 104 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |