Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198109.4 | NP_932777.2 | 10 - 11 | 983 | 68 | |
GenBank mRNA | AB099287.1 | - | 10 - 11 | 1018 | 68 | |
AK089308.1 | - | 10 - 11 | 987 | 68 | ||
AK155557.1 | - | 10 - 11 | 1008 | 68 | ||
AK165380.1 | - | 9 - 10 | 948 | 68 | ||
AK170787.1 | - | 9 - 10 | 908 | 68 | ||
AK171210.1 | - | 10 - 11 | 1032 | 68 | ||
AK171556.1 | - | 10 - 11 | 998 | 68 | ||
AK173144.1 | - | 9 - 10 | 939 | 68 | ||
BC046529.1 | - | 4 - 5 | 465 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |