Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001093754.2 | NP_001087223.1 | 2 - 3 | 462 | 90 | |
NM_001278941.1 | NP_001265870.1 | 3 - 4 | 541 | 90 | ||
NM_028110.2 | NP_082386.2 | 2 - 3 | 460 | 90 | ||
GenBank mRNA | AK008546.1 | - | 2 - 3 | 266 | 90 | |
AK050033.1 | - | 3 - 4 | 542 | 90 | ||
AK157279.1 | - | 2 - 3 | 463 | 90 | ||
BB850890.1 | - | 2 - 3 | 272 | 90 | ||
BC008266.1 | - | 2 - 3 | 450 | 2811014 | 90 | |
BC043727.1 | - | 2 - 3 | 248 | 5035764 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |