Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tusc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030254.3 | NP_084530.3 | 335 | 98 | ||
GenBank mRNA | AK028182.1 | - | 311 | 98 | ||
AK088616.1 | - | 349 | 98 | |||
AK132766.1 | - | 358 | 98 | |||
AK168664.1 | - | 293 | 98 | |||
AK168961.1 | - | 293 | 98 | |||
BC003311.1 | - | 294 | 98 | |||
BC069898.1 | - | 335 | 6489807 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |