Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175091.3 | NP_780300.2 | 23 - 24 | 3447 | 67 | |
GenBank mRNA | AK048860.1 | - | 19 - 20 | 2812 | 67 | |
BC057370.1 | - | 23 - 24 | 3428 | 6835784 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |