Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007987.2 | NP_032013.2 | 7 - 8 | 682 | 98 | |
GenBank mRNA | AK002590.1 | - | 7 - 8 | 633 | 98 | |
BC061160.1 | - | 7 - 8 | 655 | 30302649 | 98 | |
DQ846748.1 | - | 7 - 8 | 626 | 98 | ||
M83649.1 | - | 7 - 8 | 675 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |