Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hells | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008234.3 | NP_032260.2 | 14 - 15 | 1683 | 163 | |
GenBank mRNA | AF155210.1 | - | 14 - 15 | 1683 | 163 | |
AK013266.1 | - | 14 - 15 | 1722 | 163 | ||
AK147126.1 | - | 14 - 15 | 1714 | 163 | ||
AK166636.1 | - | 14 - 15 | 1684 | 163 | ||
BC090671.1 | - | 14 - 15 | 1727 | 163 | ||
U25691.2 | - | 14 - 15 | 1716 | 163 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |