Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033174.1 | NP_001028346.1 | 1 - 2 | 455 | 57 | |
GenBank mRNA | BC107327.1 | - | 1 - 2 | 189 | 57 | |
BC107328.2 | - | 1 - 2 | 189 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |