Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033174.1 | NP_001028346.1 | 6 - 7 | 1276 | 99 | |
GenBank mRNA | AK132663.1 | - | 4 - 5 | 556 | 99 | |
AK220301.1 | - | 3 - 4 | 314 | 99 | ||
BC107327.1 | - | 6 - 7 | 1010 | 99 | ||
BC107328.2 | - | 6 - 7 | 1010 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |