Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noc4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153570.2 | NP_705798.2 | 6 - 7 | 743 | 76 | |
GenBank mRNA | AK052601.1 | - | 6 - 7 | 743 | 76 | |
AK146006.1 | - | 6 - 7 | 739 | 76 | ||
AK172310.1 | - | 6 - 7 | 722 | 76 | ||
BC024616.1 | - | 6 - 7 | 733 | 4218310 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |