Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noc4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153570.2 | NP_705798.2 | 9 - 10 | 942 | 111 | |
GenBank mRNA | AK052601.1 | - | 9 - 10 | 942 | 111 | |
AK090024.1 | - | 9 - 10 | 954 | 111 | ||
AK146006.1 | - | 9 - 10 | 938 | 111 | ||
AK172310.1 | - | 9 - 10 | 921 | 111 | ||
BC024616.1 | - | 9 - 10 | 932 | 4218310 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |