Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oplah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153122.2 | NP_694762.1 | 16 - 17 | 2318 | 79 | |
GenBank mRNA | BC025120.1 | - | 3 - 4 | 416 | 4020691 | 79 |
BC034522.1 | - | 16 - 17 | 2318 | 4236444 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |