Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083188.1 | NP_001076657.1 | 6 - 7 | 734 | 112 | |
NM_001199310.1 | NP_001186239.1 | 6 - 7 | 742 | 112 | ||
NM_010715.2 | NP_034845.2 | 6 - 7 | 659 | 112 | ||
GenBank mRNA | AK153993.1 | - | 6 - 7 | 621 | 112 | |
AK168643.1 | - | 6 - 7 | 736 | 112 | ||
BC028287.1 | - | 6 - 7 | 584 | 4911261 | 112 | |
BC138541.1 | - | 6 - 7 | 742 | 8861561 | 112 | |
BC138542.1 | - | 6 - 7 | 742 | 8861562 | 112 | |
U04674.1 | - | 5 - 6 | 527 | 112 | ||
U19604.1 | - | 6 - 7 | 772 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |