Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitpnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301643.1 | NP_001288572.1 | 2 - 3 | 172 | 72 | |
NM_001301666.1 | NP_001288595.1 | 2 - 3 | 172 | 72 | ||
NM_019640.5 | NP_062614.1 | 2 - 3 | 172 | 72 | ||
GenBank mRNA | AK028235.1 | - | 2 - 3 | 107 | 72 | |
BC034676.1 | - | 2 - 3 | 62 | 4481685 | 72 | |
BQ178694.1 | - | 2 - 3 | 61 | 72 | ||
U46934.1 | - | 2 - 3 | 74 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |