Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700006969.mm.1 |
TC0700000822.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006969.mm.1 |
NM_001077591.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006969.mm.1 |
NM_001077591.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006969.mm.1 |
NM_011651.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006969.mm.1 |
NM_011651.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006966.mm.1 |
TC0700000822.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006966.mm.1 |
NM_001077591.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006966.mm.1 |
NM_001077591.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006966.mm.1 |
NM_011651.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006966.mm.1 |
NM_011651.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006967.mm.1 |
TC0700000822.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006967.mm.1 |
NM_011651.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006967.mm.1 |
NM_011651.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006968.mm.1 |
TC0700000822.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006968.mm.1 |
NM_001077591.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006968.mm.1 |
NM_001077591.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006968.mm.1 |
NM_011651.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006968.mm.1 |
NM_011651.2 |
Refseq |