Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ruvbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011304.3 | NP_035434.1 | 12 - 13 | 1323 | 76 | |
GenBank mRNA | AB013912.1 | - | 12 - 13 | 1154 | 76 | |
AK135404.1 | - | 12 - 13 | 1137 | 76 | ||
AK144156.1 | - | 12 - 13 | 1145 | 76 | ||
AK159112.1 | - | 12 - 13 | 1325 | 76 | ||
BC072585.1 | - | 1822 | 76 | |||
BC115809.1 | - | 12 - 13 | 1141 | 40105495 | 76 | |
BC116693.1 | - | 12 - 13 | 1140 | 40105493 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |