Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148927.2 | NP_683729.2 | 17 - 18 | 2030 | 113 | |
GenBank mRNA | AK010410.1 | - | 1 - 2 | 130 | 113 | |
AK088969.1 | - | 1 - 2 | 130 | 113 | ||
AV480955.1 | - | 1 - 2 | 123 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |