Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apol7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029419.2 | NP_083695.2 | 2 - 3 | 276 | 78 | |
GenBank mRNA | BC139137.1 | - | 1 - 2 | 55 | 8862159 | 78 |
BC139138.1 | - | 1 - 2 | 55 | 8862160 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |