Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kifc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010630.2 | NP_034760.2 | 1 - 2 | 124 | 123 | |
GenBank mRNA | D49545.1 | - | 1 - 2 | 200 | 123 | |
U92949.1 | - | 1 - 2 | 114 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |