Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kifc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010630.2 | NP_034760.2 | 6 - 7 | 711 | 70 | |
GenBank mRNA | AK144304.1 | - | 7 - 8 | 870 | 70 | |
D49545.1 | - | 6 - 7 | 787 | 70 | ||
U92949.1 | - | 6 - 7 | 701 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |