Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164173.1 | NP_001157645.1 | 4 - 5 | 501 | 89 | |
NM_053193.2 | NP_444423.1 | 4 - 5 | 501 | 89 | ||
GenBank mRNA | AF322193.1 | - | 4 - 5 | 472 | 89 | |
AK155648.1 | - | 4 - 5 | 442 | 89 | ||
AK155806.1 | - | 4 - 5 | 423 | 89 | ||
BC049941.1 | - | 4 - 5 | 323 | 89 | ||
BC056388.1 | - | 4 - 5 | 501 | 6854851 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |