Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankle2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253814.1 | NP_001240743.1 | 7 - 8 | 1681 | 97 | |
NM_027922.2 | NP_082198.1 | 7 - 8 | 1678 | 97 | ||
GenBank mRNA | AK003234.1 | - | 4 - 5 | 480 | 97 | |
AK129192.1 | - | 6 - 7 | 1007 | 97 | ||
AK132187.1 | - | 7 - 8 | 1683 | 97 | ||
AK142497.1 | - | 5 - 6 | 730 | 97 | ||
BC065071.1 | - | 7 - 8 | 1641 | 6410033 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |