Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195537.1 | NP_001182466.1 | 6 - 7 | 1366 | 79 | |
GenBank mRNA | AK005822.1 | - | 5 - 6 | 648 | 79 | |
BY705974.1 | - | 5 - 6 | 645 | 79 | ||
CF197814.1 | - | 4 - 5 | 442 | 79 | ||
GU144514.1 | - | 6 - 7 | 1366 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |