Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133712.2 | NP_598473.1 | 3 - 4 | 403 | 59 | |
GenBank mRNA | BC002100.1 | - | 3 - 4 | 402 | 3484608 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |