Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172685.3 | NP_766273.1 | 1 - 2 | 234 | 102 | |
GenBank mRNA | AK032806.1 | - | 1 - 2 | 235 | 102 | |
AK146034.1 | - | 1 - 2 | 233 | 102 | ||
AK167778.1 | - | 1 - 2 | 225 | 102 | ||
AK168090.1 | - | 1 - 2 | 225 | 102 | ||
AK170598.1 | - | 1 - 2 | 226 | 102 | ||
BC055369.1 | - | 1 - 2 | 223 | 30055307 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |