Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900011459.mm.1 |
NM_174857.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011459.mm.1 |
NM_174857.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011459.mm.1 |
TC1900001266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900011461.mm.1 |
NM_174857.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011461.mm.1 |
NM_174857.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011461.mm.1 |
TC1900001266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900011463.mm.1 |
NM_174857.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011463.mm.1 |
NM_174857.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011463.mm.1 |
TC1900001266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900011460.mm.1 |
NM_174857.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011460.mm.1 |
NM_174857.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011460.mm.1 |
TC1900001266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900011462.mm.1 |
NM_174857.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011462.mm.1 |
NM_174857.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011462.mm.1 |
TC1900001266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900011464.mm.1 |
TC1900001266.mm.1 |
MTA TC |