Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163144.1 | NP_001156616.1 | 1 - 2 | 680 | 101 | |
NM_001190483.1 | NP_001177412.1 | 1 - 2 | 680 | 101 | ||
GenBank mRNA | AK086210.1 | - | 1 - 2 | 665 | 101 | |
AK143998.1 | - | 1 - 2 | 681 | 101 | ||
BC013068.1 | - | 1 - 2 | 300 | 101 | ||
D12619.1 | - | 1 - 2 | 269 | 101 | ||
L14932.1 | - | 1 - 2 | 230 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus 1700 Probe spans exons |